Methyl 2,6-dibromo-3-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

38199-500MG 38199-1G 38199-5G 38199-25G
77326-640EA 87.55 USD
77326-640 77342-292 77362-930 77374-496
Methyl 2,6-dibromo-3-methoxybenzoate ≥95%
Methyl 2,6-dibromo-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈Br₂O₃
Storage Temperature: Ambient
MDL Number: MFCD32707071
CAS Number: 2383066-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR