6-Bromo-2-(4-chlorophenyl)-8-methylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

38191-500MG 38191-1G 38191-5G
77326-638EA 317.24 USD
77326-638 77350-312 77374-492
6-Bromo-2-(4-chlorophenyl)-8-methylimidazo[1,2-a]pyridine ≥97%
6-Bromo-2-(4-chlorophenyl)-8-methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrClN₂
MW: 321.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15068157
CAS Number: 910617-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR