tert-Butyl (4-bromo-2,3-dimethylphenyl)carbamate ≥97%

Supplier: AOBChem USA

Warning

38186-500MG 38186-1G 38186-5G 38186-25G
77326-636EA 144.2 USD
77326-636 77345-064 77362-926 77385-720
tert-Butyl (4-bromo-2,3-dimethylphenyl)carbamate ≥97%
tert-Butyl (4-bromo-2,3-dimethylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD29067913
CAS Number: 1385026-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR