Ethyl 2-methoxy-4,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38177-500MG 38177-1G 38177-5G 38177-25G
77326-634EA 204.97 USD
77326-634 77350-310 77362-924 77385-718
Ethyl 2-methoxy-4,5-dimethylbenzoate ≥97%
Ethyl 2-methoxy-4,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD28794954
CAS Number: 1803819-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR