Methyl (2E)-3-(2-fluorophenyl)prop-2-enoate ≥97%

Supplier: AOBChem USA

Warning

38169-1G 38169-5G 38169-25G
77340-660EA 60.77 USD
77340-660 77362-920 77374-486
Methyl (2E)-3-(2-fluorophenyl)prop-2-enoate ≥97%
Methyl (2E)-3-(2-fluorophenyl)prop-2-enoate

Precaución: for Research Use Only

Formula: C₁₀H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD03102529
CAS Number: 104201-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR