1,5-Dibromo-2-ethoxy-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

38168-500MG 38168-1G 38168-5G 38168-25G
77326-630EA 119.48 USD
77326-630 77343-646 77362-918 77374-484
1,5-Dibromo-2-ethoxy-3-methoxybenzene ≥95%
1,5-Dibromo-2-ethoxy-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32632316
CAS Number: 2384859-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR