(4,6-Dibromo-3-ethoxy-2-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38163-500MG 38163-1G 38163-5G
77326-626EA 317.24 USD
77326-626 77350-304 77374-482
(4,6-Dibromo-3-ethoxy-2-fluorophenyl)methanol ≥97%
(4,6-Dibromo-3-ethoxy-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₉Br₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD32632311
CAS Number: 2404734-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR