(4,6-Dibromo-2-fluoro-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38157-500MG 38157-1G 38157-5G 38157-25G
77326-620EA 243.08 USD
77326-620 77350-298 77362-910 77385-710
(4,6-Dibromo-2-fluoro-3-methoxyphenyl)methanol ≥97%
(4,6-Dibromo-2-fluoro-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₇Br₂FO₂
MW: 313.946 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707067
CAS Number: 2432848-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR