2-Fluoro-5-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

38152-500MG 38152-1G 38152-5G 38152-25G
77326-618EA 161.71 USD
77326-618 77345-860 77362-908 77385-708
2-Fluoro-5-methoxy-N-methylbenzamide ≥97%
2-Fluoro-5-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564062
CAS Number: 1515293-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR