2-Fluoro-5-methoxy-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

38151-500MG 38151-1G 38151-5G 38151-25G
77326-616EA 172.01 USD
77326-616 77346-416 77362-906 77385-706
2-Fluoro-5-methoxy-N,N-dimethylbenzamide ≥97%
2-Fluoro-5-methoxy-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD29091506
CAS Number: 1046468-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR