N,N-diethyl-2-fluoro-5-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

38150-500MG 38150-1G 38150-5G 38150-25G
77326-614EA 207.03 USD
77326-614 77350-296 77362-904 77385-704
N,N-diethyl-2-fluoro-5-methoxybenzamide ≥97%
N,N-diethyl-2-fluoro-5-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆FNO₂
MW: 225.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564061
CAS Number: 2734776-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR