2-Fluoro-N,5-dimethoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

38149-500MG 38149-1G 38149-5G 38149-25G
77326-612EA 126.69 USD
77326-612 77344-106 77362-902 77374-480
2-Fluoro-N,5-dimethoxy-N-methylbenzamide ≥97%
2-Fluoro-N,5-dimethoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₃
MW: 213.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28975093
CAS Number: 2385956-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR