N-Cyclopropyl-2-fluoro-5-methoxybenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
38148-500MG
38148-1G
38148-5G
38148-25G
77326-610EA
181.28
USD
77326-610
77346-834
77362-900
77385-702
N-Cyclopropyl-2-fluoro-5-methoxybenzamide ≥97%
N-Cyclopropyl-2-fluoro-5-methoxybenzamide
Precaución: for Research Use Only
Formula:
C₁₁H₁₂FNO₂ MW: 209.217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34564060 CAS Number: 1506616-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...