N-Cyclohexyl-2-fluoro-5-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

38147-500MG 38147-1G 38147-5G 38147-25G
77326-608EA 257.5 USD
77326-608 77350-294 77362-898 77385-700
N-Cyclohexyl-2-fluoro-5-methoxybenzamide ≥97%
N-Cyclohexyl-2-fluoro-5-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₂
MW: 251.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564059
CAS Number: 1543026-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR