(2-Fluoro-5-methoxyphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

38143-500MG 38143-1G 38143-5G 38143-25G
77326-602EA 136.99 USD
77326-602 77344-526 77362-892 77385-694
(2-Fluoro-5-methoxyphenyl)(thiomorpholino)methanone ≥97%
(2-Fluoro-5-methoxyphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNO₂S
MW: 255.308 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564056
CAS Number: 2734774-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR