(2-Fluoro-5-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
38142-500MG
38142-1G
38142-5G
38142-25G
77326-600EA
249.26
USD
77326-600
77350-290
77362-890
77385-692
(2-Fluoro-5-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(2-Fluoro-5-methoxyphenyl)(4-methylpiperazin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₃H₁₇FN₂O₂ MW: 252.285 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34564055 CAS Number: 2734774-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...