2-(6-Bromo-2,4-difluoro-3-methylphenyl)-1,3-dioxolane ≥95%
Supplier: AOBChem USA
|
Warning
|
38131-500MG
38131-1G
38131-5G
38131-25G
77326-598EA
93.73
USD
77326-598
77342-504
77362-888
77374-478
2-(6-Bromo-2,4-difluoro-3-methylphenyl)-1,3-dioxolane ≥95%
2-(6-Bromo-2,4-difluoro-3-methylphenyl)-1,3-dioxolane
Precaución: for Research Use Only
Formula:
C₁₀H₉BrF₂O₂ MW: 279.078 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32707061 CAS Number: 2432848-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...