1-Bromo-4-fluoro-3-isopropoxy-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

38117-500MG 38117-1G 38117-5G 38117-25G
77326-596EA 120.51 USD
77326-596 77343-688 77362-884 77374-474
1-Bromo-4-fluoro-3-isopropoxy-2-methoxybenzene ≥97%
1-Bromo-4-fluoro-3-isopropoxy-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707058
CAS Number: 2432848-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR