3-(Benzyloxy)-2-fluoro-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

38088-500MG 38088-1G 38088-25G 38088-5G
77326-594EA 297.67 USD
77326-594 77350-284 77385-690 77374-466
3-(Benzyloxy)-2-fluoro-5-(trifluoromethyl)phenol ≥97%
3-(Benzyloxy)-2-fluoro-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₄O₂
MW: 286.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632305
CAS Number: 2404734-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR