5-Bromo-2,3-difluoro-4-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

38078-500MG 38078-1G 38078-5G 38078-25G
77326-590EA 279.13 USD
77326-590 77350-282 77362-878 77385-686
5-Bromo-2,3-difluoro-4-methoxyphenol ≥97%
5-Bromo-2,3-difluoro-4-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O₂
MW: 239.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31653799
CAS Number: 1780419-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR