3-Bromo-9-propyl-9H-carbazole ≥95%

Supplier: AOBChem USA

Warning

38045-500MG 38045-1G 38045-5G 38045-25G
77326-582EA 86.52 USD
77326-582 77342-266 77362-872 77374-460
3-Bromo-9-propyl-9H-carbazole ≥95%
3-Bromo-9-propyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrN
Storage Temperature: Ambient
MDL Number: MFCD34178506
CAS Number: 865443-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR