(4-Bromo-2,6-difluorophenyl)(piperidin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
38036-500MG
38036-1G
38036-5G
38036-25G
77326-578EA
108.15
USD
77326-578
77343-166
77362-868
77374-454
(4-Bromo-2,6-difluorophenyl)(piperidin-1-yl)methanone ≥97%
(4-Bromo-2,6-difluorophenyl)(piperidin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₂H₁₂BrF₂NO Storage Temperature: Ambient |
MDL Number:
MFCD23388027 CAS Number: 1517132-49-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...