(6-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

38020-500MG 38020-1G 38020-5G 38020-25G
77326-568EA 227.63 USD
77326-568 77350-266 77362-858 77385-676
(6-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrFIS
Storage Temperature: Ambient
MDL Number: MFCD32902635
CAS Number: 2385566-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR