Methyl 2-fluoro-4-methoxy-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

38011-500MG 38011-1G 38011-5G
77339-654EA 610.79 USD
77339-654 77350-256 77374-436
Methyl 2-fluoro-4-methoxy-3-(methylthio)benzoate ≥97%
Methyl 2-fluoro-4-methoxy-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃S
MW: 230.256 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18904661
CAS Number: 1824073-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR