2-Fluoro-4-methoxy-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

38010-500MG 38010-1G 38010-5G
77335-832EA 367.71 USD
77335-832 77350-254 77374-434
2-Fluoro-4-methoxy-3-(methylthio)benzoic acid ≥97%
2-Fluoro-4-methoxy-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃S
Storage Temperature: Ambient
MDL Number: MFCD18904659
CAS Number: 1379317-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR