(3-Bromo-2-fluoro-6-methoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

38009-1G 38009-5G 38009-25G
77341-480EA 89.61 USD
77341-480 77362-854 77374-432
(3-Bromo-2-fluoro-6-methoxyphenyl)trimethylsilane ≥97%
(3-Bromo-2-fluoro-6-methoxyphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrFOSi
Storage Temperature: Ambient
MDL Number: MFCD34178505
CAS Number: 2643367-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR