2-Isopropoxy-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

37992-500MG 37992-1G 37992-5G 37992-25G
77326-560EA 211.15 USD
77326-560 77350-252 77362-850 77385-670
2-Isopropoxy-4-(trifluoromethyl)benzoic acid ≥97%
2-Isopropoxy-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD27941915
CAS Number: 1175134-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR