3,5-Difluoro-4-isopropoxyphenol ≥97%

Supplier: AOBChem USA

Warning

37988-500MG 37988-1G 37988-5G 37988-25G
77326-558EA 187.46 USD
77326-558 77350-248 77362-848 77385-668
3,5-Difluoro-4-isopropoxyphenol ≥97%
3,5-Difluoro-4-isopropoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
MW: 188.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044526
CAS Number: 1881295-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR