3,5-Difluoro-4-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37984-500MG 37984-1G 37984-5G 37984-25G
77326-554EA 202.91 USD
77326-554 77350-244 77362-844 77385-664
3,5-Difluoro-4-propoxybenzaldehyde ≥97%
3,5-Difluoro-4-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246977
CAS Number: 883521-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR