7-Methyl-2-(p-tolyl)imidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37972-500MG 37972-1G 37972-25G 37972-5G
77326-550EA 284.28 USD
77326-550 77350-240 77385-660 77374-428
7-Methyl-2-(p-tolyl)imidazo[1,2-a]pyridine ≥97%
7-Methyl-2-(p-tolyl)imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂
MW: 222.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00444716
CAS Number: 65964-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR