6-Bromo-8-methyl-2-phenylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

37968-500MG 37968-1G 37968-5G
77326-548EA 313.12 USD
77326-548 77350-238 77374-426
6-Bromo-8-methyl-2-phenylimidazo[1,2-a]pyridine ≥97%
6-Bromo-8-methyl-2-phenylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂
MW: 287.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14547277
CAS Number: 1146911-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR