1,3,5-Trichloro-2-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37954-500MG 37954-1G 37954-5G 37954-25G
77326-540EA 229.69 USD
77326-540 77350-234 77362-832 77385-654
1,3,5-Trichloro-2-propoxybenzene ≥97%
1,3,5-Trichloro-2-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉Cl₃O
MW: 239.526 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044138
CAS Number: 1881322-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR