2-Butoxy-1,3,5-trichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

37953-500MG 37953-1G 37953-5G 37953-25G
77326-538EA 210.12 USD
77326-538 77350-232 77362-830 77385-652
2-Butoxy-1,3,5-trichlorobenzene ≥97%
2-Butoxy-1,3,5-trichlorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁Cl₃O
MW: 253.553 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044160
CAS Number: 155014-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR