1,3,5-Trichloro-2-(difluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

37952-500MG 37952-1G 37952-5G 37952-25G
77326-536EA 145.23 USD
77326-536 77345-060 77362-828 77385-650
1,3,5-Trichloro-2-(difluoromethoxy)benzene ≥97%
1,3,5-Trichloro-2-(difluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃Cl₃F₂O
MW: 247.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18400134
CAS Number: 1261457-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR