5-Bromo-N,N,3-trimethylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

37932-500MG 37932-1G 37932-5G 37932-25G
77326-532EA 228.66 USD
77326-532 77350-230 77362-824 77385-646
5-Bromo-N,N,3-trimethylpicolinamide ≥97%
5-Bromo-N,N,3-trimethylpicolinamide

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂O
MW: 243.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902631
CAS Number: 2099056-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR