5-Bromo-N,N-diethyl-3-methylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

37931-500MG 37931-1G 37931-5G 37931-25G
77326-530EA 232.78 USD
77326-530 77350-228 77362-822 77385-644
5-Bromo-N,N-diethyl-3-methylpicolinamide ≥97%
5-Bromo-N,N-diethyl-3-methylpicolinamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD32902630
CAS Number: 2586127-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR