5-Bromo-N-(tert-butyl)-3-methylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

37930-500MG 37930-1G 37930-5G 37930-25G
77326-528EA 232.78 USD
77326-528 77350-226 77362-820 77385-642
5-Bromo-N-(tert-butyl)-3-methylpicolinamide ≥97%
5-Bromo-N-(tert-butyl)-3-methylpicolinamide

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD32902629
CAS Number: 156072-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR