Ethyl 2-fluoro-4,5-dimethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

37927-500MG 37927-1G 37927-5G 37927-25G
77326-524EA 141.11 USD
77326-524 77344-796 77362-816 77385-638
Ethyl 2-fluoro-4,5-dimethoxybenzoate ≥97%
Ethyl 2-fluoro-4,5-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD06204405
CAS Number: 773134-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR