2-(Benzyloxy)-4-(trifluoromethyl)phenol ≥95%

Supplier: AOBChem USA

Warning

37922-500MG 37922-1G 37922-5G
77326-522EA 325.48 USD
77326-522 77350-224 77374-420
2-(Benzyloxy)-4-(trifluoromethyl)phenol ≥95%
2-(Benzyloxy)-4-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃O₂
MW: 268.231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28973864
CAS Number: 1154743-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR