(2-(Benzyloxy)-4-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

37919-500MG 37919-1G 37919-5G 37919-25G
77326-520EA 226.6 USD
77326-520 77350-222 77362-814 77385-636
(2-(Benzyloxy)-4-(trifluoromethyl)phenyl)methanol ≥97%
(2-(Benzyloxy)-4-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃O₂
MW: 282.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27922765
CAS Number: 1823499-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR