5-Bromo-4-ethoxy-2,3-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37914-500MG 37914-1G 37914-25G 37914-5G
77326-516EA 292.52 USD
77326-516 77350-218 77385-632 77374-418
5-Bromo-4-ethoxy-2,3-difluorobenzaldehyde ≥97%
5-Bromo-4-ethoxy-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632276
CAS Number: 2404734-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR