(2-Ethoxy-3,4-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

37909-500MG 37909-1G 37909-5G
77335-494EA 356.38 USD
77335-494 77350-216 77374-416
(2-Ethoxy-3,4-difluorophenyl)methanol ≥97%
(2-Ethoxy-3,4-difluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
MW: 188.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403746
CAS Number: 2162245-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR