3,4-Difluoro-2-isopropoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

37905-500MG 37905-1G 37905-5G
77326-512EA 325.48 USD
77326-512 77350-210 77374-410
3,4-Difluoro-2-isopropoxybenzoic acid ≥95%
3,4-Difluoro-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28627607
CAS Number: 1782671-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR