5-Bromo-2,3-difluoro-4-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37898-500MG 37898-1G 37898-5G
77337-850EA 449.08 USD
77337-850 77350-204 77374-404
5-Bromo-2,3-difluoro-4-isopropoxybenzaldehyde ≥97%
5-Bromo-2,3-difluoro-4-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₂O₂
MW: 279.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902624
CAS Number: 2586126-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR