1-Bromo-3,4-difluoro-5-iodo-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

37896-500MG 37896-1G 37896-5G
77338-692EA 510.88 USD
77338-692 77350-202 77374-402
1-Bromo-3,4-difluoro-5-iodo-2-isopropoxybenzene ≥97%
1-Bromo-3,4-difluoro-5-iodo-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₂IO
MW: 376.964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902623
CAS Number: 2586126-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR