3-(5-Chloro-2,4-difluorophenyl)acrylic acid ≥97%

Supplier: AOBChem USA

Warning

37880-500MG 37880-1G 37880-5G 37880-25G
77326-510EA 151.41 USD
77326-510 77345-420 77362-810 77385-630
3-(5-Chloro-2,4-difluorophenyl)acrylic acid ≥97%
3-(5-Chloro-2,4-difluorophenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₉H₅ClF₂O₂
MW: 218.585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28750524
CAS Number: 2307843-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR