(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

37871-500MG 37871-1G 37871-5G 37871-25G
77326-508EA 222.48 USD
77326-508 77350-194 77362-808 77385-628
(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)methanol ≥97%
(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443427
CAS Number: 926190-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR