(E)-3-(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylic acid ≥97%

Supplier: AOBChem USA

Warning

37870-500MG 37870-1G 37870-5G 37870-25G
77326-506EA 264.71 USD
77326-506 77350-192 77362-806 77385-626
(E)-3-(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylic acid ≥97%
(E)-3-(7-Bromo-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉BrO₄
MW: 285.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13705206
CAS Number: 1216363-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR