(2,5-Difluoro-4-propoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

37864-500MG 37864-1G 37864-5G
77338-990EA 541.78 USD
77338-990 77350-186 77374-390
(2,5-Difluoro-4-propoxyphenyl)methanol ≥97%
(2,5-Difluoro-4-propoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756098
CAS Number: 2764728-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR