2,5-Difluoro-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37861-500MG 37861-1G 37861-5G
77338-198EA 469.68 USD
77338-198 77350-180 77374-384
2,5-Difluoro-4-propoxybenzoic acid ≥97%
2,5-Difluoro-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD32902620
CAS Number: 2586127-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR